missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIP1 Recombinant Rabbit Monoclonal Antibody (23GB6020), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | HIP1 |
|---|---|
| Clone | 23GB6020 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088854
![]() |
Invitrogen™
MA552972 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087553
|
Invitrogen™
MA552973 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HIP1 Recombinant Monoclonal antibody specifically detects HIP1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| HIP1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O00291, Q8VD75 | |
| 192154, 215114, 3092 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6020 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2610109B09Rik; A930014B11Rik; E130315I21Rik; HIP I; hip1; HIP-1; HIP-I; huntingtin interacting protein 1; huntingtin-interacting protein 1; Huntingtin-interacting protein I; ILWEQ; Kiaa4113; mKIAA4113; SHON; SHONbeta; SHONgamma | |
| A synthesized peptide derived from human HIP1 (970-1037AA). | |
| Primary | |
| -20°C | |
| HIP1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title