missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIP1 Recombinant Rabbit Monoclonal Antibody (23GB6020), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552972
This item is not returnable.
View return policy
Description
Huntingtin disease, a neurodegenerate disease, is caused by the expansion of a polymorphic glutamine tract in huntingtin. The Huntingtin Interacting Protein 1 (HIP-1) is a reportedly proapoptotic, cargo-specific adaptor protein that may be involved in the pathogenesis of Huntingtin disease. As well as playing a role in Huntingtin disease, it is likely to be involved in the recruitment of clathrin coats to lipid membranes and it may also factor in tumorigenesis by allowing the survival of precancerous and cancerous cells. Since HIP-1 expression is significantly associated with prostate and colon cancer metastasis, HIP-1 can serve as a putative prognostic factor for prostate and colon cancers.
Specifications
| HIP1 | |
| Recombinant Monoclonal | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O00291, Q8VD75 | |
| HIP1 | |
| A synthesized peptide derived from human HIP1 (970-1037AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB6020 | |
| Unconjugated | |
| HIP1 | |
| 2610109B09Rik; A930014B11Rik; E130315I21Rik; HIP I; hip1; HIP-1; HIP-I; huntingtin interacting protein 1; huntingtin-interacting protein 1; Huntingtin-interacting protein I; ILWEQ; Kiaa4113; mKIAA4113; SHON; SHONbeta; SHONgamma | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 192154, 215114, 3092 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction