missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIP1 Antibody (1B11), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
£160.00 - £368.00
Specifications
| Antigen | HIP1 |
|---|---|
| Clone | 1B11 |
| Dilution | Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation reported in scientific literature (PMID 23272104) |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18118759
|
Novus Biologicals
NB300-204SS |
0.025 mL |
£160.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18003714
|
Novus Biologicals
NB300-204 |
0.1 mL |
£368.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HIP1 Monoclonal specifically detects HIP1 in Human, Mouse, Bacteria, Rat (Negative) samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| HIP1 | |
| Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation reported in scientific literature (PMID 23272104) | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse, Bacteria, Rat (Negative) | |
| O00291 | |
| 3092 | |
| Human 3'-HIP1 (∼65 kDa) | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| 115 kDa |
| 1B11 | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| Ascites with 0.1% Sodium Azide | |
| HIP-1, HIP-I, huntingtin interacting protein 1, huntingtin-interacting protein 1, Huntingtin-interacting protein I, ILWEQ, MGC126506 | |
| HIP1 | |
| IgG1 κ | |
| Unpurified | |
| NB300-204 is specific for human HIP1. Does not cross-react with HIP1r. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title