missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIP1 Antibody (1B11), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NB300-204
This item is not returnable.
View return policy
Description
HIP1 Monoclonal specifically detects HIP1 in Human, Mouse, Bacteria, Rat (Negative) samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| HIP1 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.1% Sodium Azide | |
| HIP-1, HIP-I, huntingtin interacting protein 1, huntingtin-interacting protein 1, Huntingtin-interacting protein I, ILWEQ, MGC126506 | |
| Mouse | |
| 115 kDa | |
| 0.1 mL | |
| Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| 3092 | |
| Human, Mouse, Bacteria, Rat (Negative) | |
| Ascites |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1B11 | |
| Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation reported in scientific literature (PMID 23272104) | |
| O00291 | |
| HIP1 | |
| Human 3'-HIP1 (∼65 kDa) | |
| Unpurified | |
| RUO | |
| Primary | |
| NB300-204 is specific for human HIP1. Does not cross-react with HIP1r. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction