missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HINT2 Mouse anti-Human, Mouse, Clone: 5B10, Novus Biologicals™
Mouse Monoclonal Antibody
£250.00 - £397.00
Specifications
| Antigen | HINT2 |
|---|---|
| Clone | 5B10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18208262
|
Novus Biologicals
NBP2-59387-50UL |
50 μL |
£250.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18244093
|
Novus Biologicals
NBP2-59387-100UL |
100 μL |
£397.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HINT2 Monoclonal specifically detects HINT2 in Human samples. It is validated for Western Blot, ELISA.Specifications
| HINT2 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| EC 3.-, HINT-2, HINT-3, histidine triad nucleotide binding protein 2, histidine triad nucleotide-binding protein 2, mitochondrial, HIT-17, HIT-17kDa, PKCI-1-related HIT protein, protein kinase C inhibitor-2 | |
| HINT2 | |
| IgG1 κ | |
| Protein A purified |
| 5B10 | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 84681 | |
| Recombinant human HINT2 (18-163aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title