missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HINT2 Mouse anti-Human, Mouse, Clone: 5B10, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59387-50UL
This item is not returnable.
View return policy
Description
HINT2 Monoclonal specifically detects HINT2 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| HINT2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| EC 3.-, HINT-2, HINT-3, histidine triad nucleotide binding protein 2, histidine triad nucleotide-binding protein 2, mitochondrial, HIT-17, HIT-17kDa, PKCI-1-related HIT protein, protein kinase C inhibitor-2 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 84681 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 5B10 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| HINT2 | |
| Recombinant human HINT2 (18-163aa) purified from E. coli | |
| 50 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction