Learn More
Neogen Petrifilm™ High-Sensitivity Plate Spreader 6481
Click to view available options
Quantity:
2 Pk.
Unit Size:
Pack of 2
Description
- High-sensitivity plate spreader for Neogen Petrifilm™ High-Sensitivity Coliform Count Plates.
- Spreads inoculum over uniform area.
- Clear acrylic material allows the technician to observe and control spread.
Specifications
Specifications
| Quantity | 2 Pk. |
| Product Type | Accessory |