Learn More
Agilent BioTek High Flow Waste System for Microplate Washer
Product Code. 15517465 Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
405 LS Microplate Washer, 405 Touch Microplate Washer
Unit Size:
Each
Product Code. 15517465
Brand: Agilent BioTek 7100753S
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Accessory for 405 LS, 405 Touch microplate washers
  • For use with 405 LS , 405 Touch microplate washers
  • 4L bottles
  • 115/230V
TRUSTED_SUSTAINABILITY
Specifications
Includes 4L bottles
Voltage 115/230 V
Type High Flow Waste System
For Use With (Equipment) 405 LS Microplate Washer, 405 Touch Microplate Washer