missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HFE Rabbit anti-Human, Clone: 2O7D2, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £386.00
Specifications
| Antigen | HFE |
|---|---|
| Clone | 2O7D2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18374423
|
Novus Biologicals
NBP3-15323-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18301327
|
Novus Biologicals
NBP3-15323-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HFE Monoclonal antibody specifically detects HFE in Human samples. It is validated for Western BlotSpecifications
| HFE | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cells | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 3077 | |
| IgG | |
| Affinity purified |
| 2O7D2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| dJ221C16.10.1, hemochromatosis, hereditary hemochromatosis protein, hereditary hemochromatosis protein HLA-H, HH, high Fe, HLAH, HLA-HHFE1, MGC103790, MHC class I-like protein HFE, MVCD7 | |
| A synthetic peptide corresponding to a sequence within amino acids 249-348 of human HFE (Q30201). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title