missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HFE Rabbit anti-Human, Clone: 2O7D2, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-15323-100UL
This item is not returnable.
View return policy
Description
HFE Monoclonal antibody specifically detects HFE in Human samples. It is validated for Western Blot
Specifications
| HFE | |
| Monoclonal | |
| Unconjugated | |
| dJ221C16.10.1, hemochromatosis, hereditary hemochromatosis protein, hereditary hemochromatosis protein HLA-H, HH, high Fe, HLAH, HLA-HHFE1, MGC103790, MHC class I-like protein HFE, MVCD7 | |
| A synthetic peptide corresponding to a sequence within amino acids 249-348 of human HFE (Q30201). | |
| 100 μg | |
| Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cells | |
| 3077 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 2O7D2 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction