missing translation for 'onlineSavingsMsg'
Learn More

Hexyl nicotinate, 98%

Product Code. 15536006 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15536006

Brand: Thermo Scientific Acros 460090250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23597-82-2 | C12H17NO2 | 207.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23597-82-2
Molecular Formula C12H17NO2
Molecular Weight (g/mol) 207.27
InChI Key RVYGVBZGSFLJKH-UHFFFAOYSA-N
Synonym hexyl nicotinate, 3-pyridinecarboxylic acid, hexyl ester, hexyl nicotinoate, nicotherm, n-hexyl nicotinoate, nicotinic acid, hexyl ester, unii-bn07pb44iv, nicotinic acid n-hexyl ester, bn07pb44iv, nicotinsaeure-n-hexylester
PubChem CID 90202
IUPAC Name hexyl pyridine-3-carboxylate
SMILES CCCCCCOC(=O)C1=CN=CC=C1

Specifications

Density 1.0200g/mL
Boiling Point 329.0°C
Flash Point 128°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 25 g
Specific Gravity 1.02
Formula Weight 207.27
Percent Purity 98%
Chemical Name or Material Hexyl nicotinate
compliance-icons
Product Identifier
  • Hexyl nicotinate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.