CAS RN 23597-82-2
CAS RN 23597-82-2
IUPAC Name:
hexyl pyridine-3-carboxylate
Synonyms:
hexyl nicotinate N-hexyl nicotinoate
Molecular Weight (g/mol):
207.27
Molecular Formula:
C12H17NO2
InChi Key:
RVYGVBZGSFLJKH-UHFFFAOYSA-N
SMILES:
CCCCCCOC(=O)C1=CC=CN=C1
1
–
1
of
1
results
1
Hexyl nicotinate, 98%
CAS: 23597-82-2 Molecular Formula: C12H17NO2 Molecular Weight (g/mol): 207.27 InChI Key: RVYGVBZGSFLJKH-UHFFFAOYSA-N Synonym: hexyl nicotinate,3-pyridinecarboxylic acid, hexyl ester,hexyl nicotinoate,nicotherm,n-hexyl nicotinoate,nicotinic acid, hexyl ester,unii-bn07pb44iv,nicotinic acid n-hexyl ester,bn07pb44iv,nicotinsaeure-n-hexylester PubChem CID: 90202 IUPAC Name: hexyl pyridine-3-carboxylate SMILES: CCCCCCOC(=O)C1=CN=CC=C1