missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexokinase 2 Antibody (3D3), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
£199.00
Specifications
| Antigen | Hexokinase 2 |
|---|---|
| Clone | 3D3 |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, Knockdown Validated |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Hexokinase 2 Monoclonal specifically detects Hexokinase 2 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| Hexokinase 2 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, Knockdown Validated | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| DKFZp686M1669, EC 2.7.1, hexokinase 2, Hexokinase type II, HKII, HXK2, muscle, Muscle form hexokinase | |
| HK2 | |
| IgG1 | |
| Unpurified | |
| 102 kDa |
| 3D3 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication | |
| P52789 | |
| 3099 | |
| Purified recombinant fragment of human HK2 expressed in E. Coli (within amino acids 240-520; Uniprot # P52789). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title