missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexokinase 2 Antibody (3D3), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-51643-0.025ml
This item is not returnable.
View return policy
Description
Hexokinase 2 Monoclonal specifically detects Hexokinase 2 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications
| Hexokinase 2 | |
| Monoclonal | |
| Unconjugated | |
| P52789 | |
| HK2 | |
| Purified recombinant fragment of human HK2 expressed in E. Coli (within amino acids 240-520; Uniprot # P52789). | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
| Western Blot | |
| 3D3 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, Knockdown Validated | |
| DKFZp686M1669, EC 2.7.1, hexokinase 2, Hexokinase type II, HKII, HXK2, muscle, Muscle form hexokinase | |
| Mouse | |
| 102 kDa | |
| 0.025 mL | |
| Cell Cycle and Replication | |
| 3099 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction