missing translation for 'onlineSavingsMsg'
Learn More

Hexahelicene, TRC

Product Code. 30393672
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
5 mg
Unit Size:
0.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393672 0.5 mg 0.5mg
30407713 5 mg -
30398209 2.5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393672 Supplier Toronto Research Chemicals Supplier No. TRCH2968500.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 187-83-7
Molecular Formula C26H16
Molecular Weight (g/mol) 328.41
Synonym (±)-Hexahelicene, NSC 171474, Phenanthro[3,4-c]phenanthrene
IUPAC Name hexahelicene
SMILES c1ccc2c(c1)ccc3ccc4ccc5ccc6ccccc6c5c4c23

Specifications

Quantity 0.5 mg
InChI Formula InChI=1S/C26H16/c1-3-7-22-17(5-1)9-11-19-13-15-21-16-14-20-12-10-18-6-2-4-8-23(18)25(20)26(21)24(19)22/h1-16H
Formula Weight 328.1252
Chemical Name or Material Hexahelicene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.