CAS RN 187-83-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hexahelicene, TRC
CAS: 187-83-7 Molecular Formula: C26H16 Molecular Weight (g/mol): 328.41 Synonym: (±)-Hexahelicene,NSC 171474,Phenanthro[3,4-c]phenanthrene IUPAC Name: hexahelicene SMILES: c1ccc2c(c1)ccc3ccc4ccc5ccc6ccccc6c5c4c23