missing translation for 'onlineSavingsMsg'
Learn More

Hexaglycerin, TRC

Product Code. 30395640
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395640 1 g -
30405270 10 g 10g
30398951 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395640 Supplier Toronto Research Chemicals Supplier No. TRCH3667501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36675-34-0
Molecular Formula C18H38O13
Molecular Weight (g/mol) 462.5
SMILES C([C@@H](COC[C@@H](COC[C@H](COC[C@H](COC[C@H](COC[C@H](CO)O)O)O)O)O)O)O

Specifications

Quantity 1 g
Synonym Natrulon H 6, PCB 06, PCB 06 (polyol), Polyglycerin 06, Polyglycerin 6, Polyglycerol 6
InChI Formula InChI=1S/C18H38O13/c19-1-13(21)3-27-5-15(23)7-29-9-17(25)11-31-12-18(26)10-30-8-16(24)6-28-4-14(22)2-20/h13-26H,1-12H2/t13-,14-,15-,16-,17-,18+/m0/s1
Chemical Name or Material Hexaglycerin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.