CAS RN 36675-34-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hexaglycerin, TRC
CAS: 36675-34-0 Molecular Formula: C18H38O13 Molecular Weight (g/mol): 462.5 SMILES: C([C@@H](COC[C@@H](COC[C@H](COC[C@H](COC[C@H](COC[C@H](CO)O)O)O)O)O)O)O