missing translation for 'onlineSavingsMsg'
Learn More

Hexadecanedioic Acid, TRC

Product Code. 30389423
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389423 50 mg
30393861 10 mg
30393289 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389423 Supplier Toronto Research Chemicals Supplier No. TRCH29363350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 505-54-4
Molecular Formula C16H30O4
Molecular Weight (g/mol) 286.41
Synonym 1,14-Tetradecanedicarboxylic Acid, 1,16-Hexadecanedioic Acid, NSC 15164, Thapsic Acid, n-Tetradecane-ω,ω'-dicarboxylic Acid, alpha,ω-Tetradecanedicarboxylic Acid
IUPAC Name hexadecanedioic acid
SMILES OC(=O)CCCCCCCCCCCCCCC(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H30O4/c17-15(18)13-11-9-7-5-3-1-2-4-6-8-10-12-14-16(19)20/h1-14H2,(H,17,18)(H,19,20)
Formula Weight 286.2144
Chemical Name or Material Hexadecanedioic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.