accessibility menu, dialog, popup

UserName

HOOOHO

CAS RN 505-54-4

IUPAC Name: hexadecanedioic acid
Synonyms: hexadecanedioic acid a,w-tetradecanedicarboxylate thapsic acid thapsate
Molecular Weight (g/mol): 286.41
Molecular Formula: C16H30O4
InChi Key: QQHJDPROMQRDLA-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCCCCCCCC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)