missing translation for 'onlineSavingsMsg'
Learn More

Hexacosanoic Acid-d4, TRC

Product Code. 30396366
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396366 5 mg 5mg
30399513 10 mg 10mg
30393286 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30396366 Supplier Toronto Research Chemicals Supplier No. TRCH2909125MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1194984-85-4
Molecular Formula C26 D4 H48 O2
Molecular Weight (g/mol) 400.71
Synonym Cerotic Acid-d4, Ceratinic Acid-d4, Ceric Acid-d4, Cerinic Acid-d4, Cerylic Acid-d4, NSC 4205-d4, n-Hexacosanoic Acid-d4, Hexacosanoic-12,12,13,13-d4 Acid
IUPAC Name 12,12,13,13-tetradeuteriohexacosanoic acid
SMILES [2H]C([2H])(CCCCCCCCCCCCC)C([2H])([2H])CCCCCCCCCCC(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C26H52O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26(27)28/h2-25H2,1H3,(H,27,28)/i14D2,15D2
Formula Weight 400.4218
Chemical Name or Material Hexacosanoic Acid-d4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.