CAS RN 1194984-85-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Hexacosanoic-12,12,13,13-d4 Acid, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
Hexacosanoic Acid-d4, TRC
CAS: 1194984-85-4 Molecular Formula: C26 D4 H48 O2 Molecular Weight (g/mol): 400.71 Synonym: Cerotic Acid-d4,Ceratinic Acid-d4,Ceric Acid-d4,Cerinic Acid-d4,Cerylic Acid-d4,NSC 4205-d4,n-Hexacosanoic Acid-d4,Hexacosanoic-12,12,13,13-d4 Acid IUPAC Name: 12,12,13,13-tetradeuteriohexacosanoic acid SMILES: [2H]C([2H])(CCCCCCCCCCCCC)C([2H])([2H])CCCCCCCCCCC(=O)O