missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEXA Mouse anti-Human, Clone: 3A4F2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60337-1-IG-150UL
This item is not returnable.
View return policy
Description
This gene encodes the alpha subunit of the lysosomal enzyme beta-hexosaminidase that, together with the cofactor GM2 activator protein, catalyzes the degradation of the ganglioside GM2, and other molecules containing terminal N-acetyl hexosamines. Beta-hexosaminidase is composed of two subunits, alpha and beta, which are encoded by separate genes. Both beta-hexosaminidase alpha and beta subunits are members of family 20 of glycosyl hydrolases. Mutations in the alpha or beta subunit genes lead to an accumulation of GM2 ganglioside in neurons and neurodegenerative disorders termed the GM2 gangliosidoses. Alpha subunit gene mutations lead to Tay-Sachs disease (GM2-gangliosidosis type I).Specifications
| HEXA | |
| Monoclonal | |
| 1.86 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P06865 | |
| HEXA | |
| HEXA Fusion Protein Ag1854 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 3A4F2 | |
| Unconjugated | |
| HEXA | |
| HEXA, Hexosaminidase subunit A, TSD | |
| Mouse | |
| Protein A | |
| RUO | |
| 3073 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction