missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HES5 Recombinant Rabbit Monoclonal Antibody (23GB6360), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | HES5 |
|---|---|
| Clone | 23GB6360 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089789
![]() |
Invitrogen™
MA553026 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088658
|
Invitrogen™
MA553027 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HES5 Recombinant Monoclonal antibody specifically detects HES5 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| HES5 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P70120, Q03062, Q5TA89 | |
| 15208, 388585, 79225 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6360 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| BHLHB38; Class B basic helix-loop-helix protein 38; hairy and enhancer of split 5; hairy and enhancer of split 5 (Drosophila); hes family bHLH transcription factor 5; Hes5; Hes-5; RP3-395M20.5; Transcription factor HES-5 | |
| A synthesized peptide derived from human HES5 (1-166AA). | |
| Primary | |
| -20°C | |
| HES5 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title