missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HES5 Recombinant Rabbit Monoclonal Antibody (23GB6360), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553026
This item is not returnable.
View return policy
Description
Hairy and Enhancer-of-split homologues 5 (HES5) isa helix-loop-helix transcriptional factor that negatively regulates cell differentiation during embryogenesis. It contains one basic helix-loop-helix (bHLH) domain and one Orange domain. HES5 is reported to be a down stream effector of the Notch Signaling Pathway and is expressed in advanced ovarian serous adenocarcinomas as well as in cervical squamous carcinoma. Reports also suggest that HES5 is abundantly expressed in osteoarthritis along with Notch1 and Jagged1.
Specifications
| HES5 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| HES5 | |
| BHLHB38; Class B basic helix-loop-helix protein 38; hairy and enhancer of split 5; hairy and enhancer of split 5 (Drosophila); hes family bHLH transcription factor 5; Hes5; Hes-5; RP3-395M20.5; Transcription factor HES-5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15208, 388585, 79225 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 23GB6360 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P70120, Q03062, Q5TA89 | |
| HES5 | |
| A synthesized peptide derived from human HES5 (1-166AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction