missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HERV Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£144.00 - £397.00
Specifications
| Antigen | HERV |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:250-1:1000, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vitro assay, Blocking/Neutralizing |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In vitro Assay, Blocking Assay |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18090734
|
Novus Biologicals
NB100-93579SS |
0.025 mg |
£144.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18706653
|
Novus Biologicals
NB100-93579 |
0.1 mg |
£397.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HERV Polyclonal specifically detects HERV in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vitro assay, Block/Neutralize.Specifications
| HERV | |
| Western Blot 1:250-1:1000, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vitro assay, Blocking/Neutralizing | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q9UQF0 | |
| 30816 | |
| Synthetic peptide corresponding to residues 42-50 [QRPGNIDAPC] of the HERV protein. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 59.9 kDa |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In vitro Assay, Blocking Assay | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| endogenous retroviral family W, env(C7), member 1, ENV, envelope glycoprotein, Envelope polyprotein gPr73, enverin, Env-W, ENVW, HERV-7q, HERV7Q, HERV-7q Envelope protein, HERV-tryptophan envelope protein, HERV-W, HERV-W Env glycoprotein, HERV-W envelope protein, HERV-W_7q21.2 provirus ancestral Env polyprotein, HERV-W(7q21.1) provirus ancestral Env polyprotein, HERVWENV, HERV-W-ENV, HERVWenvelope protein, human endogenous retrovirus W envC7-1 envelope protein, Syncytin, syncytin A, Syncytin-1 | |
| ERVW-1 | |
| IgG | |
| Affinity Purified | |
| Human Endogenous Retroviruses (HERVs), a class of multiple-sclerosis-associated retroviruses |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title