missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HERV Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-93579
This item is not returnable.
View return policy
Description
HERV Polyclonal specifically detects HERV in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vitro assay, Block/Neutralize.
Specifications
| HERV | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| endogenous retroviral family W, env(C7), member 1, ENV, envelope glycoprotein, Envelope polyprotein gPr73, enverin, Env-W, ENVW, HERV-7q, HERV7Q, HERV-7q Envelope protein, HERV-tryptophan envelope protein, HERV-W, HERV-W Env glycoprotein, HERV-W envelope protein, HERV-W_7q21.2 provirus ancestral Env polyprotein, HERV-W(7q21.1) provirus ancestral Env polyprotein, HERVWENV, HERV-W-ENV, HERVWenvelope protein, human endogenous retrovirus W envC7-1 envelope protein, Syncytin, syncytin A, Syncytin-1 | |
| Rabbit | |
| 59.9 kDa | |
| 0.1 mg | |
| Primary | |
| Human Endogenous Retroviruses (HERVs), a class of multiple-sclerosis-associated retroviruses | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In vitro Assay, Blocking Assay | |
| 1 mg/mL | |
| Western Blot 1:250-1:1000, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vitro assay, Blocking/Neutralizing | |
| Q9UQF0 | |
| ERVW-1 | |
| Synthetic peptide corresponding to residues 42-50 [QRPGNIDAPC] of the HERV protein. | |
| Affinity Purified | |
| RUO | |
| 30816 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction