missing translation for 'onlineSavingsMsg'
Learn More

Herpes Simplex Virus 2 Glycoprotein D Antibody (A33) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£166.00 - £385.00

Specifications

Clone A33
Concentration 1.0 mg/mL
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Applications ELISA, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18073754
View Documents
Novus Biologicals
NB100-63170-0.025MG
0.025 mg
£166.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18049615
View Documents
Novus Biologicals
NB100-63170
0.1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Herpes Simplex Virus 2 gD Monoclonal specifically detects Herpes Simplex Virus 2 gD in Virus samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

A33
Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Monoclonal
Purified
RUO
Glycoprotein D precursor; Herpes Simplex Virus Type 2 Glycoprotein D; HHV2gp69; US6; Virion glycoprotein D
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1.0 mg/mL
ELISA, Western Blot
Unconjugated
Mouse
Virus
IgG1
Protein G purified
NB100-63170 is specific for Herpes Simplex virus 2 glycoprotein D (HSV2 gD) a major component of the virion envelope and infected cell membranes that is essential for virus entry into cells. This antibody reacts with a 56-64 kD band, corresponding to glycoprotein D, in an HSV (purified) and infected Vero cell lysate immunoblot.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.