missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herpes Simplex Virus 2 Glycoprotein D Antibody (A33) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-63170-0.025MG
This item is not returnable.
View return policy
Description
Herpes Simplex Virus 2 gD Monoclonal specifically detects Herpes Simplex Virus 2 gD in Virus samples. It is validated for Western Blot, ELISA.
Specifications
| Herpes Simplex Virus 2 gD | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| Glycoprotein D precursor; Herpes Simplex Virus Type 2 Glycoprotein D; HHV2gp69; US6; Virion glycoprotein D | |
| Herpes simplex virus, type 2, glycoprotein D | |
| 0.025 mg | |
| Primary | |
| Virus | |
| Purified |
| ELISA, Western Blot | |
| A33 | |
| Unconjugated | |
| PBS with 0.09% Sodium Azide | |
| Mouse | |
| Protein G purified | |
| RUO | |
| NB100-63170 is specific for Herpes Simplex virus 2 glycoprotein D (HSV2 gD) a major component of the virion envelope and infected cell membranes that is essential for virus entry into cells. This antibody reacts with a 56-64 kD band, corresponding to glycoprotein D, in an HSV (purified) and infected Vero cell lysate immunoblot. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction