missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HER2/ErbB2 Mouse anti-Human, Clone: 1B12A7, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | HER2/ErbB2 |
|---|---|
| Clone | 1B12A7 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16887395
|
Proteintech
60311-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16877395
|
Proteintech
60311-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This protein is a regulatory subunit of the condensin-2 complex, a complex which establishes mitotic chromosome architecture and is involved in physical rigidity of the chromatid axis.Specifications
| HER2/ErbB2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CD340, ERBB2, HER 2, HER 2/neu, HER2, MLN 19, MLN19, NEU, NGL, p185, p185erbB2, Proto oncogene c ErbB 2, Proto oncogene Neu, TKR1 | |
| ERBB2 | |
| IgG2b | |
| Protein A | |
| Antibody |
| 1B12A7 | |
| Immunohistochemistry (Paraffin), Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P04626 | |
| 2064 | |
| HER2/ErbB2 Fusion Protein Ag16463 | |
| Primary | |
| -20°C | |
| ERBB2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title