missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HER2/ErbB2 Mouse anti-Human, Clone: 1B12A7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60311-1-IG-20UL
This item is not returnable.
View return policy
Description
This protein is a regulatory subunit of the condensin-2 complex, a complex which establishes mitotic chromosome architecture and is involved in physical rigidity of the chromatid axis.Specifications
| HER2/ErbB2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P04626 | |
| ERBB2 | |
| HER2/ErbB2 Fusion Protein Ag16463 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Flow Cytometry | |
| 1B12A7 | |
| Unconjugated | |
| ERBB2 | |
| CD340, ERBB2, HER 2, HER 2/neu, HER2, MLN 19, MLN19, NEU, NGL, p185, p185erbB2, Proto oncogene c ErbB 2, Proto oncogene Neu, TKR1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 2064 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction