missing translation for 'onlineSavingsMsg'
Learn More

HER-3/c-erbB-3 Ab-2 Mouse Monoclonal Antibody, Epredia™

Epredia™ HER-3/c-erbB-3 Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

£131.00 - £531.11

Specifications

Antigen HER-3/c-erbB-3 Ab-2
Clone 2F12
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Quantity Concentration Price Quantity & Availability  
Product Code Brand Quantity Concentration Price Quantity & Availability  
12610527
View Documents
Epredia
MS-201-P0
100 μL 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12630527
View Documents
Epredia
MS-201-P1
500 μL 200μg/mL
ONLINE EXCLUSIVE PRICE
£368.00
£346.20 / 500µL
Save £21.80
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12620527
View Documents
Epredia
MS-201-P
1 mL 200μg/mL
ONLINE EXCLUSIVE PRICE
£570.00
£531.11 / 1mL
Save £38.89
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12650527
View Documents
Epredia
MS-201-P1ABX
100 μL 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12640527
View Documents
Epredia
MS-201-PABX
200 μL 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

c-erbB-3/HER-3 is the third member of the type I family of growth factor receptors. It binds to ligands in the heregulin family. c-erbB-3 is over-expressed in a variety of tumors including breast, stomach, pancreas, and colon. Heregulin & EGF stimulate tyrosine phosphorylation of c-erbB-3 to different extents.

Host Species: Mouse

Clone: 2F12

Isotype: IgG2a/κ

Species Reactivity: Human, Cow, Rat and Mouse. Others not tested.

Epitope: aa 1295-1323

Immunogen: Recombinant rat HER-3/c-erbB-3 oncoprotein

Molecular Weight: 185kDa

Positive Control: T47D cells

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

HER-3/c-erbB-3 Ab-2
Immunoprecipitation, Western Blot
Unconjugated
Mouse
Oncogenesis
Recombinant rat c-erbB-3/HER-3 oncoprotein
Primary
2F12
Monoclonal
Concentrated
RUO
Bovine, Human, Mouse, Rat
IgG2a κ
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.