Learn More
HER-3/c-erbB-3 Ab-2 Mouse Monoclonal Antibody, Epredia™
Epredia™ HER-3/c-erbB-3 Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.
Brand: Epredia MS-201-P0
Description
c-erbB-3/HER-3 is the third member of the type I family of growth factor receptors. It binds to ligands in the heregulin family. c-erbB-3 is over-expressed in a variety of tumors including breast, stomach, pancreas, and colon. Heregulin & EGF stimulate tyrosine phosphorylation of c-erbB-3 to different extents.Host Species: Mouse
Clone: 2F12
Isotype: IgG2a/κ
Species Reactivity: Human, Cow, Rat and Mouse. Others not tested.
Epitope: aa 1295-1323
Immunogen: Recombinant rat HER-3/c-erbB-3 oncoprotein
Molecular Weight: 185kDa
Positive Control: T47D cells
Cellular Localization: Cytoplasmic and membrane
Recommended for:
- Western Blotting
- Immunoprecipitation (Native and denatured)
Note:
USA: RUO; Int'l: RUO
Specifications
| HER-3/c-erbB-3 Ab-2 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 100 μL | |
| Oncogenesis | |
| Bovine, Human, Mouse, Rat | |
| IgG2a κ |
| Immunoprecipitation, Western Blot | |
| 2F12 | |
| Unconjugated | |
| Recombinant rat c-erbB-3/HER-3 oncoprotein | |
| RUO | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.