missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HER-2/c-erbB-2/neu Ab-2 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ HER-2/c-erbB-2/neu Ab-2, Mouse Monoclonal Antibody.
£120.36 - £584.00
Specifications
Antigen | HER-2/c-erbB-2/neu Ab-2 |
---|---|
Clone | 9G6.10 |
Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
12620627
|
Epredia
MS-229-P1 |
200μg/mL | 500 μL |
£347.00
500µL |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12630627
|
Epredia
MS-229-P1ABX |
1mg/mL | 100 μL |
£387.00
100µL |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12640627
|
Epredia
MS-229-PABX |
1mg/mL | 200 μL |
£584.00
200µL |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12610627
|
Epredia
MS-229-P0 |
200μg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12600627
|
Epredia
MS-229-P |
200μg/mL | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
c-erbB-2, second member (c-erbB-2/HER-2/neu) of the c-erbB family is a receptor tyrosine kinase. It exhibits extracellular domains with two cysteine-rich sequences, and a cytoplasmic tyrosine kinase domain flanked by large hydrophilic tails that carry several tyrosine autophosphorylation sites. Approximately 25% of primary breast and ovarian tumors were found to overexpress the proteinHost Species: Mouse
Clone: 9G6.10
Isotype: IgG1
Species Reactivity: Human. Does not react with mouse and rat. Others not tested.
Epitope: Extracellular domain
Immunogen: RAC311 cells transfected with SV40-neu construct, expressing human neu protein
Molecular Weight: 185kDa
Positive Control: SKBR-3 cells, or Breast CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Frozen only) Immunofluorescence
- Immunoprecipitation
- Flow Cytometry
- Inhibits Colony Formation
- Partially Inhibits Tyrosine Kinase Activity
Note:
USA: RUO; Int'l: RUO
Specifications
HER-2/c-erbB-2/neu Ab-2 | |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation | |
Unconjugated | |
Mouse | |
Oncogenesis | |
RAC311 cells transfected with SV40-neu construct, expressing human neu protein | |
Primary |
9G6.10 | |
Monoclonal | |
Concentrated | |
RUO | |
Human | |
IgG1 |