missing translation for 'onlineSavingsMsg'
Learn More

Heptanal dimethyl acetal, 96%, Thermo Scientific™

Product Code. 11336896
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11336896

Brand: Thermo Scientific Alfa Aesar L04335.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10032-05-0
Molecular Formula C9H20O2
Molecular Weight (g/mol) 160.26
MDL Number MFCD00014908
InChI Key BBMCNYFBAIUERL-UHFFFAOYSA-N
Synonym heptanal dimethyl acetal, heptane, 1,1-dimethoxy, enanthal dimethyl acetal, oenanthal dimethyl acetal, heptanal, dimethyl acetal, heptaldehyde dimethyl acetal, aldehyde c-7 dimethyl acetal, unii-rmc47kwz4j, fema no. 2541, 1,1-dimethoxy-heptane
PubChem CID 61453
IUPAC Name 1,1-dimethoxyheptane
SMILES CCCCCCC(OC)OC

Specifications

Density 0.849
Boiling Point 60°C (1 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.4115
Quantity 25 g
Beilstein 1698661
Formula Weight 160.26
Percent Purity 96%
Chemical Name or Material Heptanal dimethyl acetal
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.