accessibility menu, dialog, popup

UserName

CAS RN 10032-05-0

H3COCH3OCH3

CAS RN 10032-05-0

IUPAC Name: 1,1-dimethoxyheptane
Synonyms: heptanal dimethylacetal
Molecular Weight (g/mol): 160.26
Molecular Formula: C9H20O2
InChi Key: BBMCNYFBAIUERL-UHFFFAOYSA-N
SMILES: CCCCCCC(OC)OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results