missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Heptamethine cyanine dye-1
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | Dye Reagents |
| Molecular Weight (g/mol) | 739.17 g/mol |
| Solubility | DMSO : 50 mg/mL (67.64 mM; Need ultrasonic) |
| Content And Storage | 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light) |
| Shipping Condition | Room temperature |
| Target | HBV |
| CAS | 162411-29-2 |
| Synonym | ADS 815EI |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction