missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis C Virus Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £309.00
Specifications
| Antigen | Hepatitis C Virus |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18140923
|
Novus Biologicals
NBP2-27586SS |
0.025 mg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18116184
|
Novus Biologicals
NBP2-27586 |
0.1 mg |
£309.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hepatitis C Virus Polyclonal specifically detects Hepatitis C Virus in Virus samples. It is validated for Western Blot.Specifications
| Hepatitis C Virus | |
| Western Blot 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Virology Bacteria and Parasites | |
| PBS with 0.02% Sodium Azide | |
| Reacts with residues 33-43 [CGVYLLPRRGPR] of HCV core, env, and part of E2/NS1 of HCV. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Virus | |
| HCV NS5, Hepatitis C virus non structural protein 5, Hepatitis C virus nonstructural protein 5, Hepatitis C Virus NS5 | |
| IgG | |
| Affinity Purified | |
| Hepatitis C Virus (HCV) C3 protein |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title