missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis C Virus Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27586
This item is not returnable.
View return policy
Description
Hepatitis C Virus Polyclonal specifically detects Hepatitis C Virus in Virus samples. It is validated for Western Blot.
Specifications
| Hepatitis C Virus | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Virus | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:1000 | |
| HCV NS5, Hepatitis C virus non structural protein 5, Hepatitis C virus nonstructural protein 5, Hepatitis C Virus NS5 | |
| Reacts with residues 33-43 [CGVYLLPRRGPR] of HCV core, env, and part of E2/NS1 of HCV. | |
| 0.1 mg | |
| Virology Bacteria and Parasites | |
| Hepatitis C Virus (HCV) C3 protein | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction