missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis B Antigen Antibody (3F6-G10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-727
This item is not returnable.
View return policy
Description
Hepatitis B Antigen Monoclonal specifically detects Hepatitis B Antigen in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Hepatitis B Antigen | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:500, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 3F6-G10 | |
| Unconjugated | |
| HBV X antigen, HBx, HBxAg | |
| HB-Xag-Protein A Fusion protein | |
| 0.125 mg | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction