missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis B Antigen Antibody (3F6-G10), Novus Biologicals™
Mouse Monoclonal Antibody
£150.00 - £381.00
Specifications
| Antigen | Hepatitis B Antigen |
|---|---|
| Clone | 3F6-G10 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:500, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100 |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18209434
|
Novus Biologicals
NB600-727-0.025MG |
0.025 mg |
£150.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18285455
|
Novus Biologicals
NB600-727 |
0.125 mg |
£381.00
125µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hepatitis B Antigen Monoclonal specifically detects Hepatitis B Antigen in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Hepatitis B Antigen | |
| 1.0 mg/mL | |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Virus | |
| HB-Xag-Protein A Fusion protein | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 3F6-G10 | |
| Western Blot 1:100-1:500, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| HBV X antigen, HBx, HBxAg | |
| IgG2a | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title