missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hemoglobin Monoclonal Antibody (HB11-203.1), Invitrogen™
Mouse Monoclonal Antibody
£335.00 - £822.00
Specifications
Antigen | Hemoglobin |
---|---|
Clone | HB11-203.1 |
Concentration | 1.3 mg/mL |
Applications | ELISA, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17870196
|
Invitrogen
MA5-46006 |
100 μg |
£335.00
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11593413
|
Invitrogen
MA110807 |
500 μg |
£433.00
1mg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17800426
|
Invitrogen
MA5-46167 |
1 mg |
£822.00
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MA1-10807 detects Hemoglobin from human samples. This antibody does not detect bovine, chicken, equine, ovine, or porcine Hemoglobin. MA1-10807 has been successfully used in ELISA and Western blot applications. The MA1-10807 immunogen is purified human hemoglobin.
The human alpha globin gene cluster located on chromosome 16 spans about 30 kb and includes seven loci: 5'- zeta - pseudozeta - mu - pseudoalpha-1 - alpha-2 - alpha-1 - theta - 3'. The alpha-2 (HBA2) and alpha-1 (HBA1) coding sequences are identical. These genes differ slightly over the 5' untranslated regions and the introns, but they differ significantly over the 3' untranslated regions. Two alpha chains plus two beta chains constitute HbA, which in normal adult life comprises about 97% of the total hemoglobin; alpha chains combine with delta chains to constitute HbA-2, which with HbF (fetal hemoglobin) makes up the remaining 3% of adult hemoglobin. Alpha thalassemias result from deletions of each of the alpha genes as well as deletions of both HBA2 and HBA1; some nondeletion alpha thalassemias have also been reported.Specifications
Hemoglobin | |
1.3 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.4 | |
alpha one globin; alpha-1 globin; alpha-1-globin; alpha-2 globin chain; Alpha-globin; ALPHA-GLOBIN GENE; delta globin; Haemoglobin; Hb; HBA1; HbA1c; HBA2; HBA-T3; HBH; hemoglobin alpha 1 globin chain; Hemoglobin alpha chain; Hemoglobin subunit alpha; hemoglobin subunit alpha 1; hemoglobin, alpha 1 | |
HBA1, HBB | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
HBA1 |
HB11-203.1 | |
ELISA, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P68871, P69905 | |
3039, 3043 | |
IgG1 | |
Protein G | |
Antibody |