Learn More
Hemoglobin Monoclonal Antibody (HB11-203.1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA5-46167
Additional Details : Weight : 0.01000kg
Description
MA1-10807 detects Hemoglobin from human samples. This antibody does not detect bovine, chicken, equine, ovine, or porcine Hemoglobin. MA1-10807 has been successfully used in ELISA and Western blot applications. The MA1-10807 immunogen is purified human hemoglobin.
The human alpha globin gene cluster located on chromosome 16 spans about 30 kb and includes seven loci: 5'- zeta - pseudozeta - mu - pseudoalpha-1 - alpha-2 - alpha-1 - theta - 3'. The alpha-2 (HBA2) and alpha-1 (HBA1) coding sequences are identical. These genes differ slightly over the 5' untranslated regions and the introns, but they differ significantly over the 3' untranslated regions. Two alpha chains plus two beta chains constitute HbA, which in normal adult life comprises about 97% of the total hemoglobin; alpha chains combine with delta chains to constitute HbA-2, which with HbF (fetal hemoglobin) makes up the remaining 3% of adult hemoglobin. Alpha thalassemias result from deletions of each of the alpha genes as well as deletions of both HBA2 and HBA1; some nondeletion alpha thalassemias have also been reported.Specifications
Hemoglobin | |
Monoclonal | |
1.3 mg/mL | |
PBS with no preservative; pH 7.4 | |
P68871, P69905 | |
HBA1, HBB | |
Purified human hemoglobin. | |
1 mg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
HB11-203.1 | |
Unconjugated | |
HBB | |
alpha one globin; alpha-1 globin; alpha-1-globin; alpha-2 globin chain; Alpha-globin; ALPHA-GLOBIN GENE; delta globin; Haemoglobin; Hb; HBA1; HbA1c; HBA2; HBA-T3; HBH; hemoglobin alpha 1 globin chain; Hemoglobin alpha chain; Hemoglobin subunit alpha; hemoglobin subunit alpha 1; hemoglobin, alpha 1 | |
Mouse | |
Protein G | |
RUO | |
3039, 3043 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |