missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEMK2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56486
This item is not returnable.
View return policy
Description
HEMK2 Polyclonal specifically detects HEMK2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| HEMK2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| C21orf127, chromosome 21 open reading frame 127, EC 2.1.1.-, HemK methyltransferase family member 2, HEMK2, M.HsaHemK2P, MGC19995, MTQ2, N(6)-adenine-specific DNA methyltransferase 1, N-6 adenine-specific DNA methyltransferase 1 (putative), N6AMT, N6-DNA-methyltransferase, PRED28 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Equine: 100%; Rat: 100%; Bovine: 92%; Canine: 92%; Mouse: 92%. | |
| Human, Mouse, Rat, Bovine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q96F73 | |
| N6AMT1 | |
| Synthetic peptides corresponding to N6AMT1(N-6 adenine-specific DNA methyltransferase 1 (putative)) The peptide sequence was selected from the N terminal of N6AMT1. Peptide sequence MAGENFATPFHGHVGRGAFSDVYEPAEDTFLLLNALEAAAAELAGVEICL. | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 29104 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction