missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HELLS Recombinant Rabbit Monoclonal Antibody (4M7J3)

Rabbit Recombinant Monoclonal Antibody
£147.00 - £363.00
Specifications
| Antigen | HELLS |
|---|---|
| Clone | 4M7J3 |
| Concentration | 1.78 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30283945
|
Invitrogen™
MA558029 |
20 μL |
£147.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30282996
![]() |
Invitrogen™
MA558030 |
100 μL |
£363.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: RGHKVLLFSQ MTSMLDILMD YCHLRDFNFS RLDGSMSYSE REKNMHSFNT DPEVFIFLVS TRAGGLGINL TAADTVIIYD SDWNPQSDLQ AQDRCHRIGQ TKPVVVYRLV TANTIDQKIV ERAAAKRKLE KLIIHKNHFK GGQSGLNLSK NFLDPKELME LLKSRDYERE IKGSREKVIS DKDLELLLDR SDLIDQMNAS GPIKEKMGIF KILENSEDSS PECLF.
This gene encodes a lymphoid-specific helicase. Other helicases function in processes involving DNA strand separation, including replication, repair, recombination, and transcription. This protein is thought to be involved with cellular proliferation and may play a role in leukemogenesis. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.Specifications
| HELLS | |
| 1.78 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q9NRZ9 | |
| 3070 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 614-838 of human Lsh/HELLS (NP_060533.2). | |
| Antibody |
| 4M7J3 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| HELLS | |
| AI323785; E130115I21Rik; helicase, lymphoid specific; helicase, lymphoid-specific; hells; ICF4; LSH; Lymphocyte-specific helicase; lymphoid-specific helicase; Lysh; Nbla10143; PASG; Proliferation-associated SNF2-like protein; SMARCA6; SWI/SNF2-related matrix-associated actin-dependent regulator of chromatin subfamily A member 6; SWI/SNF2-related, matrix-associated, actin-dependent regulator of chromatin, subfamily A, member 6; Tbc1d12; TBC1D12: TBC1 domain family, member 12; YFK8 | |
| HELLS | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title