missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HELLS Recombinant Rabbit Monoclonal Antibody (4M7J3)

Product Code. 30283945
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30283945 20 μL 20µL
30282996 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30283945 Supplier Invitrogen™ Supplier No. MA558029

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: RGHKVLLFSQ MTSMLDILMD YCHLRDFNFS RLDGSMSYSE REKNMHSFNT DPEVFIFLVS TRAGGLGINL TAADTVIIYD SDWNPQSDLQ AQDRCHRIGQ TKPVVVYRLV TANTIDQKIV ERAAAKRKLE KLIIHKNHFK GGQSGLNLSK NFLDPKELME LLKSRDYERE IKGSREKVIS DKDLELLLDR SDLIDQMNAS GPIKEKMGIF KILENSEDSS PECLF.

This gene encodes a lymphoid-specific helicase. Other helicases function in processes involving DNA strand separation, including replication, repair, recombination, and transcription. This protein is thought to be involved with cellular proliferation and may play a role in leukemogenesis. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HELLS
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 4M7J3
Concentration 1.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Gene HELLS
Gene Accession No. Q9NRZ9
Gene Alias AI323785; E130115I21Rik; helicase, lymphoid specific; helicase, lymphoid-specific; hells; ICF4; LSH; Lymphocyte-specific helicase; lymphoid-specific helicase; Lysh; Nbla10143; PASG; Proliferation-associated SNF2-like protein; SMARCA6; SWI/SNF2-related matrix-associated actin-dependent regulator of chromatin subfamily A member 6; SWI/SNF2-related, matrix-associated, actin-dependent regulator of chromatin, subfamily A, member 6; Tbc1d12; TBC1D12: TBC1 domain family, member 12; YFK8
Gene Symbols HELLS
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 614-838 of human Lsh/HELLS (NP_060533.2).
Purification Method Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3070
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.