missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HELLS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | HELLS |
|---|---|
| Concentration | 0.3 mg/mL |
| Applications | Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16899363
|
Proteintech
11955-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16889363
|
Proteintech
11955-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the polycystin protein family. The encoded protein contains multiple transmembrane domains, and cytoplasmic N- and C-termini. The protein may be an integral membrane protein involved in cell-cell/matrix interactions. The encoded protein may function in renal tubular development, morphology, and function, and may modulate intracellular calcium homoeostasis and other signal transduction pathways. This protein interacts with polycystin 1 to produce cation-permeable currents. Mutations in this gene have been associated with autosomal domit polycystic kidney disease.Specifications
| HELLS | |
| Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| Q60848, Q9NRZ9 | |
| 15201, 294071, 3070 | |
| HELLS Fusion Protein Ag2558 | |
| Primary | |
| -20°C | |
| HELLS |
| 0.3 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| helicase, lymphoid, HELLS, LSH, Lymphoid specific helicase, Nbla10143, PASG, SMARCA6 | |
| Hells | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title