missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HELLS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11955-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the polycystin protein family. The encoded protein contains multiple transmembrane domains, and cytoplasmic N- and C-termini. The protein may be an integral membrane protein involved in cell-cell/matrix interactions. The encoded protein may function in renal tubular development, morphology, and function, and may modulate intracellular calcium homoeostasis and other signal transduction pathways. This protein interacts with polycystin 1 to produce cation-permeable currents. Mutations in this gene have been associated with autosomal domit polycystic kidney disease.Specifications
| HELLS | |
| Polyclonal | |
| Unconjugated | |
| HELLS | |
| helicase, lymphoid, HELLS, LSH, Lymphoid specific helicase, Nbla10143, PASG, SMARCA6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15201, 294071, 3070 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q60848, Q9NRZ9 | |
| Hells | |
| HELLS Fusion Protein Ag2558 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction