Learn More
Thermo Scientific™ Hektoen Enteric Agar
Description
Isolate and differentiate Shigella and Salmonella spp. from clinical specimens and food, with this selective medium.
Isolate and differentiate Shigella and Salmonella spp. from clinical specimens and food, with ready-to-use Thermo Scientific™ Oxoid™ Hektoen Enteric Agar. This selective medium is recommended for the isolation of salmonella from foods according to the Bacteriological Analytical Manual1. It is also recommended with the ISO 215672 standards for the isolation of shigellae from foods.
Hektoen Enteric Agar is a differential and selective medium for the isolation of Shigella and Salmonella spp.
- Recommended for the isolation of salmonella from foods according to the Bacteriological Analytical Manual1.
- Conforms to ISO 21567 for isolation of shigellae from foods2.
- Easy-to-read: shigellae form green moist raised colonies, while salmonellae form blue green colonies, and other coliforms form salmon-pink to orange colonies surrounded by a zone of bile precipitation.
- Ready-to-use convenience of prepared media.
The high peptone content in the medium offsets the inhibitory effects of bile salts on Shigella species in particular. The additional carbohydrates (sucrose and salicin) give better differentiation than lactose alone and the lower toxicity of the double indicator improves recovery. The increased lactose content helps early recognition of slow lactose-fermenting organisms. The thiosulphate and ferric citrate are present to detect H2S producing organisms.
Specifications
Specifications
| Color | Olive to ochre yellow |
| Content And Storage | 2°C to 10°C |
| Format | 90mm Monoplate |
| Manufacturing Origin | Germany |
| Product Type | Agar Plate |
| Fill Weight | 17g ± 5% |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.