missing translation for 'onlineSavingsMsg'
Learn More

HEK 293T cells Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen HEK 293T cells
Concentration 1.33 mg/mL
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16825695
View Documents
Proteintech
27555-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16815695
View Documents
Proteintech
27555-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody is designed to detect the proteins of the HEK 293T cells.

HEK 293T cell line, an important variant of HEK 293 cell line, which contains the SV40 Large T-antigen that allows for episomal replication of transfected plasmids containing the SV40 origin of replication. HEK 293T cells are usually used for plasmids transfection and temporal expression of desired gene products.
Specifications

Specifications

HEK 293T cells
ELISA
Unconjugated
Rabbit
Human
293T, HEK 293T cells, HEK293T
Primary
-20°C
1.33 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
IgG
Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.